Learn More
Biomatik Corporation Human CD81 (CD81 antigen) ELISA Kit
Supplier:  Biomatik Corporation EKF58938
Encompass_Preferred
Human CD81 (CD81 antigen) ELISA Kit for detecting CD81 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P60033. Target Synonyms: CD81; CD81 antigen; CD81 molecule; S5.7; TAPA1; Tetraspanin 28; Tspan 28; TSPAN28. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8215