Learn More
Biomatik Corporation Human Myoglobin (MYO) ELISA Kit, 96 tests
Catalog No. 501503313
Encompass_Preferred
The Human Myoglobin (MYO) ELISA Kit allows for quantitative determination of Myoglobin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02144. Cross Activity: No significant cross-reactivity or interference between Myoglobin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3313 Supplier Biomatik Corporation Supplier No. EKU06078
The Human Myoglobin (MYO) ELISA Kit allows for quantitative determination of Myoglobin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02144. Cross Activity: No significant cross-reactivity or interference between Myoglobin and analogues was observed. Restriction: For Research Use Only.