Learn More
Biomatik Corporation Human FGG (Fibrinogen gamma chain) ELISA Kit
Supplier:  Biomatik Corporation EKF59043
Encompass_Preferred
Human FGG (Fibrinogen gamma chain) ELISA Kit for detecting FGG in plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.469-30ng/ml. Sensitivity: 0.281ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02679. Target Synonyms: FGγ; Fibrinogen Gamma chain; FGG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8232