Learn More
Biomatik Corporation Rat Myoglobin (MYO) ELISA Kit, 96 tests
Catalog No. 501503315
Encompass_Preferred
Rat Myoglobin (MYO) ELISA Kit for detecting MYO in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.277ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QZ76. Gene ID: 59108. Target Synonyms: PVALB; MB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3315 Supplier Biomatik Corporation Supplier No. EKU06081
Rat Myoglobin (MYO) ELISA Kit for detecting MYO in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.277ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QZ76. Gene ID: 59108. Target Synonyms: PVALB; MB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.