Learn More
Biomatik Corporation Human Myosin Binding Protein C, Cardiac (MYBPC3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06085
Encompass_Preferred
The Human Myosin Binding Protein C, Cardiac (MYBPC3) ELISA Kit allows for quantitative determination of Myosin Binding Protein C, Cardiac in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.063 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q14896. Cross Activity: No significant cross-reactivity or interference between Myosin Binding Protein C, Cardiac and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3316