Learn More
Biomatik Corporation Rat GT (Gastrin) ELISA Kit
Supplier:  Biomatik Corporation EKF59158
Encompass_Preferred
Rat GT (Gastrin) ELISA Kit for detecting GT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20pg/ml. Sensitivity: 0.188pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P04563. Target Synonyms: Gastrin; Gastrin-34; G34; Gast; Gas; GT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8245