Learn More
Biomatik Corporation Human SBDP 145 (Alpha II SPECTRIN BREAKDOWN PRODUCT 145) ELISA Kit
Supplier:  Biomatik Corporation EKF59167
Encompass_Preferred
Human SBDP 145 (Alpha II SPECTRIN BREAKDOWN PRODUCT 145) ELISA Kit for detecting SBDP 145 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q13813. Target Synonyms: SBDP145; SBDP-145. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8247