Learn More
Biomatik Corporation Human Myosin Heavy Chain 9, Non Muscle (MYH9) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06104
Encompass_Preferred
The Human Myosin Heavy Chain 9, Non Muscle (MYH9) ELISA Kit allows for quantitative determination of Myosin Heavy Chain 9, Non Muscle in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P35579. Cross Activity: No significant cross-reactivity or interference between Myosin Heavy Chain 9, Non Muscle and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3321