Learn More
Biomatik Corporation Human AQP-4 (Aquaporin-4) ELISA Kit
Supplier:  Biomatik Corporation EKF59205
Encompass_Preferred
Human AQP-4 (Aquaporin-4) ELISA Kit for detecting AQP-4 in cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P55087. Target Synonyms: AQP-4; Aquaporin-4; AQP4; HMIWC2; MIWC; WCH4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8251