Learn More
Biomatik Corporation Human Myosin Light Chain 1, Alkali, Fast Skeletal (MYL1) ELISA Kit, 96 tests
Catalog No. 501503322
Encompass_Preferred
The Human Myosin Light Chain 1, Alkali, Fast Skeletal (MYL1) ELISA Kit allows for quantitative determination of Myosin Light Chain 1, Alkali, Fast Skeletal in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05976. Cross Activity: No significant cross-reactivity or interference between Myosin Light Chain 1, Alkali, Fast Skeletal and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3322 Supplier Biomatik Corporation Supplier No. EKU06105
The Human Myosin Light Chain 1, Alkali, Fast Skeletal (MYL1) ELISA Kit allows for quantitative determination of Myosin Light Chain 1, Alkali, Fast Skeletal in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05976. Cross Activity: No significant cross-reactivity or interference between Myosin Light Chain 1, Alkali, Fast Skeletal and analogues was observed. Restriction: For Research Use Only.