Learn More
Biomatik Corporation Human TPSAB1 (Tryptase alpha/beta-1) ELISA Kit
Supplier:  Biomatik Corporation EKF59210
Encompass_Preferred
Human TPSAB1 (Tryptase alpha/beta-1) ELISA Kit for detecting TPSAB1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q15661. Target Synonyms: Tryptase alpha; TPS1; TPSAB1; Tryptase 1; TPS2; TPSB1; tryptase alpha; beta 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8253