Learn More
Biomatik Corporation Chicken IgA (Immunoglobulin A) ELISA Kit
Supplier:  Biomatik Corporation EKF59287
Encompass_Preferred
Chicken IgA (Immunoglobulin A) ELISA Kit for detecting IgA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Target Synonyms: IgA; Immunoglobulin A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8259