Learn More
Biomatik Corporation Human N-Acetyl Alpha-D-Glucosaminidase (NAGLU) ELISA Kit, 96 tests
Catalog No. 501503327
Encompass_Preferred
The Human N-Acetyl Alpha-D-Glucosaminidase (NAGLU) ELISA Kit allows for quantitative determination of N-Acetyl Alpha-D-Glucosaminidase in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.113 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P54802. Cross Activity: No significant cross-reactivity or interference between N-Acetyl Alpha-D-Glucosaminidase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3327 Supplier Biomatik Corporation Supplier No. EKU06128
The Human N-Acetyl Alpha-D-Glucosaminidase (NAGLU) ELISA Kit allows for quantitative determination of N-Acetyl Alpha-D-Glucosaminidase in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.113 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P54802. Cross Activity: No significant cross-reactivity or interference between N-Acetyl Alpha-D-Glucosaminidase and analogues was observed. Restriction: For Research Use Only.