Learn More
Biomatik Corporation Human N-Acetyl Beta-D-Glucosaminidase (NAGase) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06129
Encompass_Preferred
The Human N-Acetyl Beta-D-Glucosaminidase (NAGase) ELISA Kit allows for quantitative determination of N-Acetyl Beta-D-Glucosaminidase in Urine. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.54 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O60502. Cross Activity: No significant cross-reactivity or interference between N-Acetyl Beta-D-Glucosaminidase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3328