Learn More
Biomatik Corporation Hamster IFN-γ (Interferon gamma) ELISA Kit
Supplier:  Biomatik Corporation EKF59543
Encompass_Preferred
Hamster IFN-γ (Interferon gamma) ELISA Kit for detecting IFN-γ in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Hamster. Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O35497. Target Synonyms: IFN-γ; IFNG; IFG; IFI; Type II Interferon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8273