Learn More
Biomatik Corporation Porcine D2D (D-Dimer) ELISA Kit
Supplier:  Biomatik Corporation EKF59684
Encompass_Preferred
Porcine D2D (D-Dimer) ELISA Kit for detecting D2D in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: F1RX36; A0A5G2R564; I3LJW2. Target Synonyms: D-Dimer; D2D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8281