Learn More
Biomatik Corporation Mouse Nephrin (NPHN) ELISA Kit, 96 tests
Catalog No. 501503333
Encompass_Preferred
Mouse Nephrin (NPHN) ELISA Kit for detecting NPHN in urine, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QZS7. Gene ID: 54631. Target Synonyms: NPHS1; CNF; Nephrosis 1; Congenital Finnish Type; Renal glomerulus-specific cell adhesion receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3333 Supplier Biomatik Corporation Supplier No. EKU06164
Mouse Nephrin (NPHN) ELISA Kit for detecting NPHN in urine, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QZS7. Gene ID: 54631. Target Synonyms: NPHS1; CNF; Nephrosis 1; Congenital Finnish Type; Renal glomerulus-specific cell adhesion receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.