Learn More
Biomatik Corporation Spermine ELISA Kit
Supplier:  Biomatik Corporation EKF59995
Encompass_Preferred
Spermine ELISA Kit for detecting Spermine in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 12.5-800ng/ml. Sensitivity: 7.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: Spermine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8290