Learn More
Biomatik Corporation Rat Nephrin (NPHN) ELISA Kit, 96 tests
Catalog No. 501503334
Encompass_Preferred
Rat Nephrin (NPHN) ELISA Kit for detecting NPHN in urine, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9R044. Gene ID: 64563. Target Synonyms: NPHS1; CNF; Nephrosis 1; Congenital Finnish Type; Renal glomerulus-specific cell adhesion receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3334 Supplier Biomatik Corporation Supplier No. EKU06165
Rat Nephrin (NPHN) ELISA Kit for detecting NPHN in urine, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9R044. Gene ID: 64563. Target Synonyms: NPHS1; CNF; Nephrosis 1; Congenital Finnish Type; Renal glomerulus-specific cell adhesion receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.