Learn More
Biomatik Corporation Mouse Nephronectin (NPNT) ELISA Kit, 96 tests
Catalog No. 501503335
Encompass_Preferred
Mouse Nephronectin (NPNT) ELISA Kit for detecting NPNT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91V88. Gene ID: 114249. Target Synonyms: EGFL6L; POEM; Preosteoblast EGF-like repeat protein with MAM domain; Protein EGFL6-like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3335 Supplier Biomatik Corporation Supplier No. EKU06167
Mouse Nephronectin (NPNT) ELISA Kit for detecting NPNT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91V88. Gene ID: 114249. Target Synonyms: EGFL6L; POEM; Preosteoblast EGF-like repeat protein with MAM domain; Protein EGFL6-like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.