Learn More
Biomatik Corporation Human Neprilysin (NEP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06168
Encompass_Preferred
The Human Neprilysin (NEP) ELISA Kit allows for quantitative determination of Neprilysin in Serum, Plasma, cerebrospinal fluid and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 11.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08473. Cross Activity: No significant cross-reactivity or interference between Neprilysin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3336