Learn More
Biomatik Corporation LPS (Lipopolysaccharides) ELISA Kit
Supplier:  Biomatik Corporation EKF60245
Encompass_Preferred
LPS (Lipopolysaccharides) ELISA Kit for detecting LPS in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Universal. Detection Range: 0.313-20ug/ml. Sensitivity: 0.188ug/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antibody. Assay Time: 3h, 30 min. Target Synonyms: LPS; Lipopolysaccharides. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8297