Learn More
Biomatik Corporation Mouse Neprilysin (NEP) ELISA Kit, 96 tests
Catalog No. 501503337
Encompass_Preferred
Mouse Neprilysin (CD10) ELISA Kit for detecting CD10 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61391. Gene ID: 17380. Target Synonyms: NEP; MME; CALLA; SFE; Atriopeptidase; Enkephalinase; Skin fibroblast elastase; Common Acute Lymphoblastic Leukemia Antigen; Membrane Metallo-Endopeptidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3337 Supplier Biomatik Corporation Supplier No. EKU06169
Mouse Neprilysin (CD10) ELISA Kit for detecting CD10 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61391. Gene ID: 17380. Target Synonyms: NEP; MME; CALLA; SFE; Atriopeptidase; Enkephalinase; Skin fibroblast elastase; Common Acute Lymphoblastic Leukemia Antigen; Membrane Metallo-Endopeptidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.