Learn More
Biomatik Corporation Rat Adcyap1 (Pituitary adenylate cyclase-activating polypeptide) ELISA Kit
Supplier:  Biomatik Corporation EKF60369
Encompass_Preferred
Rat Adcyap1 (Pituitary adenylate cyclase-activating polypeptide) ELISA Kit for detecting Adcyap1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400pg/ml. Sensitivity: 3.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P13589. Target Synonyms: Adcyap1; PACAP; Pituitary adenylate cyclase-activating polypeptide; ADCYAP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8306