Learn More
Biomatik Corporation Mouse Nerve Growth Factor (NGF) ELISA Kit, 96 tests
Catalog No. 501503340
Encompass_Preferred
Mouse Nerve Growth Factor (NGF) ELISA Kit for detecting NGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6LDU8. Gene ID: 18049. Target Synonyms: NGFB; Beta-NGF; HSAN5; NGF-B; Beta-Nerve Growth Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3340 Supplier Biomatik Corporation Supplier No. EKU06175
Mouse Nerve Growth Factor (NGF) ELISA Kit for detecting NGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6LDU8. Gene ID: 18049. Target Synonyms: NGFB; Beta-NGF; HSAN5; NGF-B; Beta-Nerve Growth Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.