Learn More
Biomatik Corporation Rat PACAP-38 (Pituitary Adenylate Cyclase Activating Polypeptide 38) ELISA Kit
Supplier:  Biomatik Corporation EKF60375
Encompass_Preferred
Rat PACAP-38 (Pituitary Adenylate Cyclase Activating Polypeptide 38) ELISA Kit for detecting PACAP-38 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 125-8000pg/ml. Sensitivity: 75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P13589. Target Synonyms: PACAP-38. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8312