Learn More
Biomatik Corporation Human Nesfatin 1 (NES1) ELISA Kit, 96 tests
Catalog No. 501503342
Encompass_Preferred
The Human Nesfatin 1 (NES1) ELISA Kit allows for quantitative determination of Nesfatin 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 617.3-50000 pg/mL. Sensitivity: 234.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P80303. Cross Activity: No significant cross-reactivity or interference between Nesfatin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3342 Supplier Biomatik Corporation Supplier No. EKU06180
The Human Nesfatin 1 (NES1) ELISA Kit allows for quantitative determination of Nesfatin 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 617.3-50000 pg/mL. Sensitivity: 234.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P80303. Cross Activity: No significant cross-reactivity or interference between Nesfatin 1 and analogues was observed. Restriction: For Research Use Only.