Learn More
Biomatik Corporation Human ENG (Endoglin) ELISA Kit
Supplier:  Biomatik Corporation EKF60381
Encompass_Preferred
Human ENG (Endoglin) ELISA Kit for detecting ENG in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P17813. Target Synonyms: Endoglin; CD105; ENG; END; HHT1; ORW; ORW1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8318