Learn More
Biomatik Corporation Mouse Nestin (NES) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06185
Encompass_Preferred
Mouse Nestin (NES) ELISA Kit for detecting NES in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6P5H2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3343