Learn More
Biomatik Corporation Human PKD1 (Polycystin-1) ELISA Kit
Supplier:  Biomatik Corporation EKF60387
Encompass_Preferred
Human PKD1 (Polycystin-1) ELISA Kit for detecting PKD1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P98161. Target Synonyms: PKD1; Polycystic kidney disease protein 1-like 1; PC1-like 1 protein; Polycystin-1L1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8323