Learn More
Biomatik Corporation Human Netrin 4 (Ntn4) ELISA Kit, 96 tests
Catalog No. 501503346
Encompass_Preferred
The Human Netrin 4 (Ntn4) ELISA Kit allows for quantitative determination of Netrin 4 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9HB63. Cross Activity: No significant cross-reactivity or interference between Netrin 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3346 Supplier Biomatik Corporation Supplier No. EKU06191
The Human Netrin 4 (Ntn4) ELISA Kit allows for quantitative determination of Netrin 4 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9HB63. Cross Activity: No significant cross-reactivity or interference between Netrin 4 and analogues was observed. Restriction: For Research Use Only.