Learn More
Biomatik Corporation Mouse Neuregulin 1 (NRG1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06197
Encompass_Preferred
Mouse Neuregulin 1 (NRG1) ELISA Kit for detecting NRG1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6DR98. Target Synonyms: ARIA; GGF; HGL; HRG; HRGA; NDF; SMDF; Acetylcholine receptor-inducing activity; Glial growth factor; Heregulin; Neu differentiation factor; Sensory and motor neuron-derived factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3347