Learn More
Biomatik Corporation Human IAPP (Islet amyloid polypeptide) ELISA Kit
Supplier:  Biomatik Corporation EKF60397
Encompass_Preferred
Human IAPP (Islet amyloid polypeptide) ELISA Kit for detecting IAPP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P10997. Target Synonyms: IAPP; DAP; IAP; Amylin; Islet amyloid polypeptide; Insulinoma amyloid peptide; Diabetes-associated peptide; DAP;. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8331