Learn More
Biomatik Corporation Human Neurofilament, Heavy Polypeptide (NEFH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06213
Encompass_Preferred
The Human Neurofilament, Heavy Polypeptide (NEFH) ELISA Kit allows for quantitative determination of Neurofilament, Heavy Polypeptide in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P12036. Cross Activity: No significant cross-reactivity or interference between Neurofilament, Heavy Polypeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3350