Learn More
Biomatik Corporation Rat Neurofilament, Heavy Polypeptide (NEFH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06214
Encompass_Preferred
Rat Neurofilament, Heavy Polypeptide (NEFH) ELISA Kit for detecting NEFH in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16884. Target Synonyms: NFH; 200 kDa neurofilament protein; Neurofilament triplet H protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3351