Learn More
Biomatik Corporation Mouse IgG (Immunoglobulin G) ELISA Kit
Supplier:  Biomatik Corporation EKF60445
Encompass_Preferred
Mouse IgG (Immunoglobulin G) ELISA Kit for detecting IgG in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01867; P01868. Target Synonyms: IgG; Immunoglobulin G. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8354