Learn More
Biomatik Corporation Human Neuroglobin (NGB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06219
Encompass_Preferred
The Human Neuroglobin (NGB) ELISA Kit allows for quantitative determination of Neuroglobin in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 2.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NPG2. Cross Activity: No significant cross-reactivity or interference between Neuroglobin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3354