Learn More
Biomatik Corporation Human Neuropeptide FF (NPFF) ELISA Kit, 96 tests
Catalog No. 501503358
Encompass_Preferred
The Human Neuropeptide FF (NPFF) ELISA Kit allows for quantitative determination of Neuropeptide FF in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-1000 pg/mL. Sensitivity: 26.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: O15130. Cross Activity: No significant cross-reactivity or interference between Neuropeptide FF and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3358 Supplier Biomatik Corporation Supplier No. EKU06241
The Human Neuropeptide FF (NPFF) ELISA Kit allows for quantitative determination of Neuropeptide FF in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-1000 pg/mL. Sensitivity: 26.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: O15130. Cross Activity: No significant cross-reactivity or interference between Neuropeptide FF and analogues was observed. Restriction: For Research Use Only.