Learn More
Biomatik Corporation Human Neuropeptide S (NPS) ELISA Kit, 96 tests
Catalog No. 501503360
Encompass_Preferred
The Human Neuropeptide S (NPS) ELISA Kit allows for quantitative determination of Neuropeptide S in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1,000 pg/mL. Sensitivity: 4.48 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P0C0P6. Cross Activity: No significant cross-reactivity or interference between Neuropeptide S and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3360 Supplier Biomatik Corporation Supplier No. EKU06243
The Human Neuropeptide S (NPS) ELISA Kit allows for quantitative determination of Neuropeptide S in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.35-1,000 pg/mL. Sensitivity: 4.48 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P0C0P6. Cross Activity: No significant cross-reactivity or interference between Neuropeptide S and analogues was observed. Restriction: For Research Use Only.