Learn More
Biomatik Corporation Human Neuropeptide Y (NPY) ELISA Kit, 96 tests
Catalog No. 501503361
Encompass_Preferred
The Human Neuropeptide Y (NPY) ELISA Kit allows for quantitative determination of Neuropeptide Y in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 24.69-2000 pg/mL. Sensitivity: 9.18 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01303. Cross Activity: No significant cross-reactivity or interference between Neuropeptide Y and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3361 Supplier Biomatik Corporation Supplier No. EKU06247
Add to Cart
Add to Cart
The Human Neuropeptide Y (NPY) ELISA Kit allows for quantitative determination of Neuropeptide Y in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 24.69-2000 pg/mL. Sensitivity: 9.18 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P01303. Cross Activity: No significant cross-reactivity or interference between Neuropeptide Y and analogues was observed. Restriction: For Research Use Only.