Learn More
Biomatik Corporation Human Neurotensin (NT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06265
Encompass_Preferred
The Human Neurotensin (NT) ELISA Kit allows for quantitative determination of Neurotensin in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 37.0-3,000 pg/mL. Sensitivity: 13.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P30990. Cross Activity: No significant cross-reactivity or interference between Neurotensin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3365