Learn More
Biomatik Corporation Human Neurotrophin 4 (NT4) ELISA Kit, 96 tests
Catalog No. 501503368
Encompass_Preferred
The Human Neurotrophin 4 (NT4) ELISA Kit allows for quantitative determination of Neurotrophin 4 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.0 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P34130. Cross Activity: No significant cross-reactivity or interference between Neurotrophin 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3368 Supplier Biomatik Corporation Supplier No. EKU06273
The Human Neurotrophin 4 (NT4) ELISA Kit allows for quantitative determination of Neurotrophin 4 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.0 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P34130. Cross Activity: No significant cross-reactivity or interference between Neurotrophin 4 and analogues was observed. Restriction: For Research Use Only.