Learn More
Biomatik Corporation Human Neutrophil Activating Protein 3 (NAP3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06279
Encompass_Preferred
The Human Neutrophil Activating Protein 3 (NAP3) ELISA Kit allows for quantitative determination of Neutrophil Activating Protein 3 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P09341. Cross Activity: No significant cross-reactivity or interference between Neutrophil Activating Protein 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3369