Learn More
Biomatik Corporation Mouse Neutrophil Activating Protein 3 (NAP3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06280
Encompass_Preferred
Mouse Chemokine (C-X-C Motif) Ligand 1 (CXCL1) ELISA Kit for detecting C-X-C Motif in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12850. Gene ID: 14825. Target Synonyms: NAP3; GRO1; GROa; GRO-A; MGSA-A; NAP3; SCYB1; Chemokine C-X-C-Motif Ligand 1; Melanoma Growth Stimulating Activity Alpha; Fibroblast Secretory Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3370