Learn More
Biomatik Corporation Chicken IL-8 (Interleukin 8) ELISA Kit
Supplier:  Biomatik Corporation EKF60745
Encompass_Preferred
Chicken IL-8 (Interleukin 8) ELISA Kit for detecting IL-8 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08317. Target Synonyms: IL-8; IL8; CXCL8; GCP-1; GCP1; LECT; LUCT; LYNAP; MDNCF; MONAP; NAF; NAP-1; NAP1; 3-10C; AMCF-I; K60. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8420