Learn More
Biomatik Corporation Mouse Neutrophil Gelatinase Associated Lipocalin (NGAL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06285
Encompass_Preferred
Mouse Neutrophil gelatinase-associated lipocalin (NGAL) ELISA Kit for detecting NGAL in serum, plasma, tissue homogenates, cell lysates, urine, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11672. Gene ID: 16819. Target Synonyms: Lipocalin-2; LCN2; p25; Lipocalin 2; Oncogene 24p3; 25 kDa alpha-2-microglobulin-related subunit of MMP-9; Siderocalin LCN2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3372