Learn More
Biomatik Corporation Human GzmB (Granzyme B) ELISA Kit
Supplier:  Biomatik Corporation EKF60752
Encompass_Preferred
Human GzmB (Granzyme B) ELISA Kit for detecting GzmB in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100pg/ml. Sensitivity: 0.938pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P10144. Target Synonyms: GzmB; Granzyme B; Gzmb; CSPB; CTLA-1; CTSGL1; Fragmentin-2; Granzyme-2; GRB; GrzB; GZMB; HLP; SECT; Granzyme B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8424