Learn More
Biomatik Corporation Rat Nicotinamide Adenine Dinucleotide Phosphate Oxidase 1 (NOX1) ELISA Kit, 96 tests
Catalog No. 501503375
Encompass_Preferred
Rat Nicotinamide Adenine Dinucleotide Phosphate Oxidase 1 (NOX1) ELISA Kit for detecting NOX1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.56ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WV87. Gene ID: 114243. Target Synonyms: MOX1; GP91-2; NOH1; NADPH Oxidase 1; Mitogenic Oxidase (Pyridine Nucleotide-Dependent Superoxide-Generating; NADH/NADPH mitogenic oxidase subunit P65-MOX. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3375 Supplier Biomatik Corporation Supplier No. EKU06292
Rat Nicotinamide Adenine Dinucleotide Phosphate Oxidase 1 (NOX1) ELISA Kit for detecting NOX1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.56ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WV87. Gene ID: 114243. Target Synonyms: MOX1; GP91-2; NOH1; NADPH Oxidase 1; Mitogenic Oxidase (Pyridine Nucleotide-Dependent Superoxide-Generating; NADH/NADPH mitogenic oxidase subunit P65-MOX. Restrictions: For Research Use Only. Not for use in diagnostic procedures.