Learn More
Biomatik Corporation Mouse Nicotinamide Adenine Dinucleotide Phosphate Oxidase 4 (NOX4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06294
Encompass_Preferred
Mouse NADPH Oxidase 4 (NOX4) ELISA Kit for detecting NOX4 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JHI8. Gene ID: 50490. Target Synonyms: KOX; KOX-1; RENOX; Nicotinamide Adenine Dinucleotide Phosphate Oxidase 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3377