Learn More
Biomatik Corporation Human Niemann Pick Disease Type C2 (NPC2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU06302
Encompass_Preferred
The Human Niemann Pick Disease Type C2 (NPC2) ELISA Kit allows for quantitative determination of Niemann Pick Disease Type C2 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.29 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P61916. Cross Activity: No significant cross-reactivity or interference between Niemann Pick Disease Type C2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3378